missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL5 Antibody (Z88), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54809JF549
This item is not returnable.
View return policy
Description
ELOVL5 Monoclonal specifically detects ELOVL5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| ELOVL5 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ELOVL5 | |
| Peptide Sequence NNVKPRKLR | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Z88 | |
| Western Blot, ELISA, Immunohistochemistry | |
| dJ483K16.1, EC 2.3.1.n8, elongation of very long chain fatty acids protein 5, ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, ELOVL2,3-keto acyl-CoA synthase ELOVL5, Fatty acid elongase 1, hELO1, HELO1homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2, homolog of yeast long chain polyunsaturated fatty acid elongatio, SUR4/Elo3-like, yeast) | |
| Mouse | |
| Protein A purified | |
| Cardiovascular Biology, Cellular Signaling | |
| 60481 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction