missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35681-100ul
This item is not returnable.
View return policy
Description
ELOVL5 Polyclonal antibody specifically detects ELOVL5 in Human samples. It is validated for ELISA,Western Blot
Specifications
| ELOVL5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| dJ483K16.1, EC 2.3.1.n8, elongation of very long chain fatty acids protein 5, ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, ELOVL2,3-keto acyl-CoA synthase ELOVL5, Fatty acid elongase 1, hELO1, HELO1homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2, homolog of yeast long chain polyunsaturated fatty acid elongatio, SUR4/Elo3-like, yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 247-326 of human ELOVL5 (NP_001229757.1).,, Sequence:, GVIWPCTFPLGWLYFQIGYMISLIALFTNFYIQTYNKKGASRRKDHLKDHQNGSMAAVNGHTNSFSPLENNVKPRKLRKD | |
| 100 μL | |
| Cardiovascular Biology, Cellular Signaling | |
| 60481 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction