missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£176.00 - £385.00
Specifications
| Antigen | ELOVL4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18304946
|
Novus Biologicals
NBP3-03895-20UL |
20 μL |
£176.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18333216
|
Novus Biologicals
NBP3-03895-100UL |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELOVL4 Polyclonal specifically detects ELOVL4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ELOVL4 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ADMD, cancer/testis antigen 118, CT118, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4, elongation of very long chain fatty acids protein 4, FLJ17667, FLJ92876,3-keto acyl-CoA synthase ELOVL4, Stargardt disease 3 (autosomal dominant), STGD2, STGD3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ELOVL4 (NP_073563.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Vision | |
| PBS with 50% glycerol, pH7.3. | |
| 6785 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title