missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £401.00
Specifications
| Antigen | ELOVL4 |
|---|---|
| Dilution | Western Blot 1:400, Immunohistochemistry 10 - 15 ug/ml |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18317761
|
Bio-Techne
NBP3-11920-25UG |
25 μg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18385873
|
Novus Biologicals
NBP3-11920-100UG |
100 μg |
£401.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELOVL4 Polyclonal antibody specifically detects ELOVL4 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| ELOVL4 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Vision | |
| PBS | |
| 6785 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot 1:400, Immunohistochemistry 10 - 15 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ADMD, cancer/testis antigen 118, CT118, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4, elongation of very long chain fatty acids protein 4, FLJ17667, FLJ92876,3-keto acyl-CoA synthase ELOVL4, Stargardt disease 3 (autosomal dominant), STGD2, STGD3 | |
| Synthetic peptdide derived from the human ELOVL4 protein (Uniprot: Q9GZR5) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title