missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELOVL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599281
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ELOVL4.
This gene encodes a membrane-bound protein which is a member of the ELO family, proteins which participate in the biosynthesis of fatty acids. Consistent with the expression of the encoded protein in photoreceptor cells of the retina, mutations and small deletions in this gene are associated with Stargardt-like macular dystrophy (STGD3) and autosomal dominant Stargardt-like macular dystrophy (ADMD), also referred to as autosomal dominant atrophic macular degeneration.
Specifications
| ELOVL4 | |
| Polyclonal | |
| Unconjugated | |
| ELOVL4 | |
| 3-keto acyl-CoA synthase Elovl4; ADMD; cancer/testis antigen 118; CT118; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4; elongation of very long chain fatty acids protein 4; elongation of very long chain fatty acids-like 4; ELOVL FA elongase 4; ELOVL fatty acid elongase 4; Elovl4; FLJ17667; FLJ92876; ISQMR; SCA34; STGD2; STGD3; Very long chain 3-ketoacyl-CoA synthase 4; very long chain 3-oxoacyl-CoA synthase 4; very-long-chain 3-oxoacyl-CoA synthase 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6785, 83603 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9EQC4, Q9GZR5 | |
| ELOVL4 | |
| A synthesized peptide derived from human ELOVL4(Accession Q9GZR5), corresponding to amino acid residues S45-R95. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction