missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELOVL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550027
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ELOVL3 protein.
This gene encodes a protein that belongs to the GNS1/SUR4 family. Members of this family play a role in elongation of long chain fatty acids to provide precursors for synthesis of sphingolipids and ceramides. [provided by RefSeq, Jul 2013].
Specifications
| ELOVL3 | |
| Polyclonal | |
| Unconjugated | |
| ELOVL3 | |
| 3-keto acyl-CoA synthase ELOVL3; CIG30; CIG-30; Cold-inducible glycoprotein of 30 kDa; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3; elongation of very long chain fatty acids protein 3; ELOVL FA elongase 3; ELOVL fatty acid elongase 3; ELOVL3; Very long chain 3-ketoacyl-CoA synthase 3; very long chain 3-oxoacyl-CoA synthase 3; very-long-chain 3-oxoacyl-CoA synthase 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83401 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HB03 | |
| ELOVL3 | |
| Synthesized peptide derived from internal of human ELOVL3 (aa 31-80). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction