missing translation for 'onlineSavingsMsg'
Learn More

ELOVL3 Antibody (V61P2B3-D10), Novus Biologicals™

Product Code. 18351161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351161 0.1 mg 0.10mg
18624829 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18351161 Supplier Novus Biologicals Supplier No. NBP276673

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ELOVL3 Monoclonal specifically detects ELOVL3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELOVL3
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V61P2B3-D10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA, Immunohistochemistry
Formulation PBS pH 7.4, 50% glycerol with 0.09% sodium azide
Gene Alias 3-keto acyl-CoA synthase ELOVL3, CIG-30, CIG30MGC21435, Cold-inducible glycoprotein of 30 kDa, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3, elongation of very long chain fatty acids protein 3
Gene Symbols ELOVL3
Host Species Mouse
Immunogen Synthetic peptide of Human Elongation of very long chain fatty acids protein 3
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 83401
Test Specificity Detects ∼40 kDa
Target Species Human, Rat
Content And Storage Store at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.