missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL3 Antibody (V61 P2B3*D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £315.00
Specifications
| Antigen | ELOVL3 |
|---|---|
| Clone | V61 P2B3*D10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668846
|
Novus Biologicals
NBP2-50385-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668207
|
Novus Biologicals
NBP2-50385 |
0.1 mL |
£315.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELOVL3 Monoclonal antibody specifically detects ELOVL3 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| ELOVL3 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Signaling, metabolism | |
| PBS | |
| 83401 | |
| IgG1 κ | |
| Protein A purified |
| V61 P2B3*D10 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 3-keto acyl-CoA synthase ELOVL3, CIG-30, CIG30MGC21435, Cold-inducible glycoprotein of 30 kDa, EC 2.3.1.n8, elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3, elongation of very long chain fatty acids protein 3 | |
| Ovalbumin-conjugated synthetic peptide - QPYNFELSK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title