missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELOA2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110467

Product Code. 17148393

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AQVLRNNPDA LSDVGEVPYW VLEPVLEGWR PDQLYRRKKD NHALVRETDE LRRNHCFQDF KEEKPQENKT WREQYLRLPD APEQRLRVMT TNIRSARGNN PNGREAKMIC FKSVAKTPYD TSRRQEKSAG DADPENGEIK PASKPAGSSH TPSSQSSSGG GRDSSSSILR WLPEKRANPC LSSSNEHAAP AAKTRKQAAK KVAPLMAKAI RDYKRRFSRR.

This gene encodes the transcriptionally active subunit of the SIII (or elongin) transcription elongation factor complex, which also includes two regulatory subunits, elongins B and C. This complex acts to increase the rate of RNA chain elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites along the DNA template. Whereas a related protein with similar function, elongin A, is ubiquitously expressed, the encoded protein is specifically expressed in the testis, suggesting it may have a role in spermatogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELOA2
Polyclonal
Unconjugated
ELOA2
EloA2; Elongin-A2; HsT832; RNA polymerase II transcription factor SIII subunit A2; TCEB3B; TCEB3L; transcription elongation factor (SIII) elongin A2 elongin A2; transcription elongation factor B polypeptide 3B; transcription elongation factor B subunit 3B
Rabbit
Affinity Chromatography
RUO
51224
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.68 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IYF1
ELOA2
Recombinant fusion protein containing a sequence corresponding to amino acids 534-753 of human TCEB3B (NP_057511.2).
100 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.