missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELOA Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £341.00
Specifications
| Antigen | ELOA |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17919242
![]() |
Invitrogen™
PA5145399 |
200 μL |
£341.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17939612
|
Invitrogen™
PA5145400 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes the transcriptionally active subunit of the SIII (or elongin) transcription elongation factor complex, which also includes two regulatory subunits, elongins B and C. This complex acts to increase the rate of RNA chain elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites along the DNA template. Whereas a related protein with similar function, elongin A, is ubiquitously expressed, the encoded protein is specifically expressed in the testis, suggesting it may have a role in spermatogenesis.Specifications
| ELOA | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q14241, Q8CB77, Q8IYF1 | |
| 27224, 51224, 6924 | |
| Recombinant Mouse ELOA protein, Val76-Ser345 (Accession #Q8CB77). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 110kDa; AA408125; El; EloA; EloA2; Elongin 110 kDa subunit; elongin A; Elongin A2; elongin-A; Elongin-A2; MSTP059; RNA polymerase II transcription factor elongin subunit A (110 kDa); RNA polymerase II transcription factor SIII subunit A; RNA polymerase II transcription factor SIII subunit A1; SIII; SIII p110; TCEB3; TCEB3A; TCEB3B; TCEB3L; transcription elongation factor B (SIII), polypeptide 3; transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A); transcription elongation factor B alpha subunit; transcription elongation factor B polypeptide 3; transcription elongation factor B subunit 3 | |
| ELOA, ELOA2 | |
| IgG | |
| Antigen affinity chromatography, Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title