missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELOA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550839
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Elongin A protein.
This gene encodes the transcriptionally active subunit of the SIII (or elongin) transcription elongation factor complex, which also includes two regulatory subunits, elongins B and C. This complex acts to increase the rate of RNA chain elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites along the DNA template. Whereas a related protein with similar function, elongin A, is ubiquitously expressed, the encoded protein is specifically expressed in the testis, suggesting it may have a role in spermatogenesis.
Specifications
| ELOA | |
| Polyclonal | |
| Unconjugated | |
| ELOA | |
| 110kDa; AA408125; El; EloA; EloA2; Elongin 110 kDa subunit; elongin A; Elongin A2; elongin-A; Elongin-A2; MSTP059; RNA polymerase II transcription factor elongin subunit A (110 kDa); RNA polymerase II transcription factor SIII subunit A; RNA polymerase II transcription factor SIII subunit A1; SIII; SIII p110; TCEB3; TCEB3A; TCEB3B; TCEB3L; transcription elongation factor B (SIII), polypeptide 3; transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A); transcription elongation factor B alpha subunit; transcription elongation factor B polypeptide 3; transcription elongation factor B subunit 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51224, 6924 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q14241, Q8IYF1 | |
| ELOA, ELOA2 | |
| Synthetic peptide corresponding to a region derived from internal residues of human transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction