missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELMSAN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570873
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 85%; Pig: 92%; Rabbit: 85%; Rat: 93%.
ELMSAN1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleus; DNA binding.
Specifications
| ELMSAN1 | |
| Polyclonal | |
| Unconjugated | |
| ELMSAN1 | |
| 9430029N19Rik; C130039O16Rik; c14_5541; C14orf117; C14orf43; C8H14orf43; ELM2 and Myb/SANT domain containing 1; ELM2 and Myb/SANT-like domain containing 1; ELM2 and SANT domain-containing protein 1; ELMSAN1; Gm260; LOW QUALITY PROTEIN: ELM2 and SANT domain-containing protein 1; LSR68; MIDEAS; mitotic deacetylase associated SANT domain protein; RGD1306119 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 91748 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6PJG2 | |
| ELMSAN1 | |
| synthetic peptide directed towards the C-terminal region of human ELMSAN1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction