missing translation for 'onlineSavingsMsg'
Learn More

ELMSAN1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18408804 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408804 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408804 Supplier Novus Biologicals Supplier No. NBP171912UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ELMSAN1 Polyclonal antibody specifically detects ELMSAN1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMSAN1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias c14_5541, C14orf117, C14orf43, chromosome 14 open reading frame 117, chromosome 14 open reading frame 43, ELM2 and Myb/SANT-like domain containing 1, ELMSAN1, FLJ00335, hypothetical protein LOC91748, LSR68
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 550 and 600 of human ELMSAN1 Open Reading Frame 43 using the numbering given in entry NP_001036783.1 (GeneID 91748).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91748
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.