missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELMOD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596711
This item is not returnable.
View return policy
Description
Immunogen sequence: ATFLHLAHVW RTQRKTISDS GFVLKELEVL AKKSPRRLLK TLELYLARVS KGQASLLGAQ KCYGPEAPPF KDLTFTGESD LQSHSSEGVW LI; Positive Samples: A-549, Mouse brain, Rat brain; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, kinocilium, stereocilium.
RBM29 (RNA binding motif protein 29), also known as ELMOD3 (ELMO domain-containing protein 3) or RBED1 (RNA-binding motif and ELMO domain-containing protein 1), is a 381 amino acid protein that contains one ELMO domain and exists as six alternatively spliced isoforms. The gene encoding RBM29 maps to human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
Specifications
| ELMOD3 | |
| Polyclonal | |
| Unconjugated | |
| ELMOD3 | |
| AI844780; C330008I15Rik; deafness, autosomal recessive 88; DFNB88; ELMO domain containing 3; ELMO domain-containing protein 3; ELMO/CED-12 domain containing 3; Elmod3; Kcmf1; liver-specific organic anion transporter 3TM12; LST3; organic anion transporter LST-3b; potassium channel modulatory factor 1; PP4068; Rbed1; RBM29; RNA binding motif and ELMO domain 1; RNA binding motif and ELMO/CED-12 domain 1; RNA binding motif protein 29; RNA-binding motif and ELMO domain-containing protein 1; RNA-binding motif protein 29; RNA-binding protein 29 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 232089, 297342, 84173 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.35 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5XIQ2, Q91YP6, Q96FG2 | |
| ELMOD3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-381 of human ELMOD3 (NP_0011284931). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction