missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ELMOD2 Polyclonal specifically detects ELMOD2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ELMOD2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | ELMO domain containing 2, ELMO domain-containing protein 2,9830169G11Rik, ELMO/CED-12 domain containing 2, FLJ35768, FLJ38038, MGC10084 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein of mouse ELMOD2 |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?