Learn More
Invitrogen™ ELMO3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596602
Description
Immunogen sequence: NKTWKEMRAT QEDFDKVMQV VREQLARTLA LKPTSLELFR TKVNALTYGE VLRLRQTERL HQEGTLAPPI LELREKLKPE LMGLIRQQRL LRLCEGTLFR KISSRRRQDK LWFCCLSPNH KLLQYGDMEE GASPPTLESL PEQLPVADMR ALLTGKDCPH VREKGSGKQN KDLYELAFSI SYDRGEEEAY LNFIAPSKRE FYLWTDGLSA LLGSPMGSEQ TRLDLEQLLT METKLRLLEL ENVPIPERPP PVPPPPTNFN FCYDCSIAEP; Positive Samples: 22Rv1, SW480, HT-29, Mouse brain, Mouse liver, Mouse kidney, Rat lung; Cellular Location: Cytoplasm.
The protein encoded by this gene is similar to a C. elegans protein that functions in phagocytosis of apoptotic cells and in cell migration. Other members of this small family of engulfment and cell motility (ELMO) proteins have been shown to interact with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes.
Specifications
| ELMO3 | |
| Polyclonal | |
| Unconjugated | |
| ELMO3 | |
| 9930107J06; BC058752; Ced12; CED-12; ced-12 homolog 3; Elmo3; ELMO-3; engulfment and cell motility 3; engulfment and cell motility 3, ced-12 homolog; engulfment and cell motility protein 3; FLJ13824 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 234683, 291962, 79767 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q499U2, Q8BYZ7, Q96BJ8 | |
| ELMO3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 504-773 of human ELMO3 (NP_0789882). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.