missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELMO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530792
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (84%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein. Similarity to a C. elegans protein suggests that this protein may function in phagocytosis of apoptotic cells and in cell migration. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| ELMO2 | |
| Polyclonal | |
| Unconjugated | |
| ELMO2 | |
| 1190002F24Rik; CED12; CED-12; ced-12 homolog 2; CED-12 homolog A; CED12A; Ced-12A; Elmo2; ELMO-2; engulfment and cell motility 2; engulfment and cell motility 2, ced-12 homolog; engulfment and cell motility protein 2; FLJ11656; hCed-12A; KIAA1834; PH domain protein CED12A; protein ced-12 homolog A; RP11-394O2.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63916 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96JJ3 | |
| ELMO2 | |
| Recombinant protein encompassing a sequence within the center region of human ELMO2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction