missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELMO1 Recombinant Rabbit Monoclonal Antibody (JG34-59)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534729
This item is not returnable.
View return policy
Description
Positive Control: Rat spleen tissue, mouse spleen tissue, Jurkat, SH-SY-5Y.
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. Similarity to a C. elegans protein suggests that this protein may function in apoptosis and in cell migration. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
Specifications
| ELMO1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q92556 | |
| ELMO1 | |
| Synthetic peptide within Human ELMO1 aa 678-727. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Western Blot | |
| JG34-59 | |
| Unconjugated | |
| ELMO1 | |
| 6330578D22Rik; C230095H21Rik; CED12; CED-12; ced-12 homolog 1; ELMO1; ELMO-1; ELMO1s; engulfment and cell motility 1; engulfment and cell motility 1 (ced-12 homolog, C. elegans); engulfment and cell motility 1 isoform 2; engulfment and cell motility 1 splice 1; engulfment and cell motility 1, ced-12 homolog; engulfment and cell motility protein 1; im:7151147; KIAA0281; MGC126406; OTTHUMP00000207056; OTTHUMP00000207060; Protein ced-12 homolog; wu:fq24e08; zgc:85612 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9844 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction