missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELMO1 Recombinant Rabbit Monoclonal Antibody (9X6Y6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538138
This item is not returnable.
View return policy
Description
Positive Samples: Raji, SH-SY5Y, Mouse lung, Mouse brain, Rat brain Immunogen sequence: KGALKQNKEV LELAFSILYD SNCQLNFIAP DKHEYCIWTD GLNALLGKDM MSDLTRNDLD TLLSMEIKLR LLDLENIQIP DAPPPIPKEP SNYDFVYDCN.
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. Similarity to a C. elegans protein suggests that this protein may function in apoptosis and in cell migration. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
Specifications
| ELMO1 | |
| Recombinant Monoclonal | |
| 0.37 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BPU7, Q92556 | |
| ELMO1 | |
| A synthetic peptide corresponding to a sequence within amino acids 628-727 of human ELMO1 (Q92556). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 9X6Y6 | |
| Unconjugated | |
| ELMO1 | |
| 6330578D22Rik; C230095H21Rik; CED12; CED-12; ced-12 homolog 1; ELMO1; ELMO-1; ELMO1s; engulfment and cell motility 1; engulfment and cell motility 1 (ced-12 homolog, C. elegans); engulfment and cell motility 1 isoform 2; engulfment and cell motility 1 splice 1; engulfment and cell motility 1, ced-12 homolog; engulfment and cell motility protein 1; im:7151147; KIAA0281; MGC126406; OTTHUMP00000207056; OTTHUMP00000207060; Protein ced-12 homolog; wu:fq24e08; zgc:85612 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140580, 361251, 9844 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction