missing translation for 'onlineSavingsMsg'
Learn More

ELMO1 Rabbit anti-Human, Mouse, Rat, Clone: 9X6Y6, Novus Biologicals™

Product Code. 18333677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333677 100 μg 100µL
18310686 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18333677 Supplier Novus Biologicals Supplier No. NBP316410100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ELMO1 Monoclonal antibody specifically detects ELMO1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMO1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9X6Y6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ced-12 homolog 1, CED12CED-12, ELMO-1, engulfment and cell motility 1, engulfment and cell motility 1 (ced-12 homolog, C. elegans), engulfment and cell motility protein 1, KIAA0281MGC126406, Protein ced-12 homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 628-727 of human ELMO1 (Q92556).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9844
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.