Learn More
Elma Elmasonic Easy 60 H
Product Code. 17874505
Change view
Click to view available options
Plug Type:
UK-Plug
Unit Size:
Each
Product Code. Plug Type unitSize
17874505 UK-Plug Each
1 options
Product Code. 17874505 Supplier Elma Supplier No. 1118894003
Add to basket
Add to basket
This water bath includes 9 units of different sizes and is characterized by simple, user-friendly operation. With 100% ultrasonic power at a frequency of 37 kHz, it solves cleaning tasks quickly and efficiently. Suited to remove tenacious, mineral soiling and polishing pastes.
  • 37 kHz ultrasonic frequency (control range -2.5 / +5.5 kHz)
  • 2 ultrasonic modes for need-based sound field distribution in the ultrasonic bath
  • Electronic selector switches on wipeable membrane keypad and LED display of all operating states
  • Pre-selection of cleaning time: cleaning time adjustable between 1 minute and 30 minutes or continuous operation
  • Electronic temperature control adjustable in steps of 5
  • Safety switch-off with wake-up function - after 8 hours of inactivity - in case of a brief interruption of the power supply
  • Automatic shutdown of ultrasonic operation
  • Splash-proof housing and cleaning tray made of stainless steel
  • Side stopcock for emptying the cleaning tank via the drain at the rear of the unit
  • 2-year warranty from the initial operation, but no longer than 3 years from the production date
Specifications
Display None
Material Stainless Steel
Timer 1 to 30 min. and continuous
Frequency 50/60 Hz
Voltage 220/240 V
Capacity (English) 199.5 oz.
Capacity (Metric) 5.9 L
Dimensions (D x W x H) Interior 130 x 280 x 145 mm
Includes Lid
Model Easy 60 H (with heating)
Plug Type UK-Plug