missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ellis van Creveld syndrome Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Ellis van Creveld syndrome Polyclonal specifically detects Ellis van Creveld syndrome in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Ellis van Creveld syndrome |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | Ellis van Creveld syndrome, ellis-van Creveld syndrome protein |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 743-992 of human Ellis van Creveld syndrome (NP_714928.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?