missing translation for 'onlineSavingsMsg'
Learn More

Ellagic acid, MP Biomedicals™

Product Code. 11416842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416842 10 g 10g
11496832 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416842 Supplier MP Biomedicals Supplier No. 0215102910
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Color Brown-Yellow
Quantity 10 g
Formula Weight 302.2
Percent Purity 0.999
Physical Form Powder
Chemical Name or Material Ellagic acid, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.