missing translation for 'onlineSavingsMsg'
Learn More

Ellagic acid, MedChemExpress

Product Code. 16422079
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422079 1 g 1g
16410808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422079 Supplier Medchem Express Supplier No. HYB0183
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ellagic acid is a natural antioxidant, and acts as a potent and ATP-competitive CK2 inhibitor, with an IC50 of 40 nM and a Ki of 20 nM.

Ellagic acid is a natural antioxidant, and acts as a potent and ATP-competitive CK2 inhibitor, with an IC50 of 40 nM and a Ki of 20 nM.

Specifications

Chemical Name or Material Ellagic acid
CAS 476-66-4
Color Light Nude
Purity Grade Notes Research
Molecular Formula C14H6O8
Quantity 1 g
Solubility Information DMSO : 5 mg/mL (16.55 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : < 1 mg/mL (insoluble) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C2=CC(O)=C(O)C(O3)=C2C4=C(C3=O)C=C(O)C(O)=C4O1
Molecular Weight (g/mol) 302.19
Formula Weight 302.19
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.