missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ELL2 Polyclonal specifically detects ELL2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ELL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | ELL-related RNA polymerase II, elongation factor, elongation factor, RNA polymerase II, 2, RNA polymerase II elongation factor ELL2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 253-352 of human ELL2 (NP_036213.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?