missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELL Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | ELL |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18302214
|
Novus Biologicals
NBP3-03945-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352054
|
Novus Biologicals
NBP3-03945-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELL Polyclonal specifically detects ELL in Mouse samples. It is validated for Western Blot.Specifications
| ELL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 8178 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| C19orf17, chromosome 19 open reading frame 17, DKFZp434I1916, Eleven-nineteen lysine-rich leukemia protein, ELL gene (11-19 lysine-rich leukemia gene), ELL1, elongation factor RNA polymerase II, MEN, RNA polymerase II elongation factor ELL | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human ELL (NP_006523.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title