missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELL Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81668JF549
This item is not returnable.
View return policy
Description
ELL Polyclonal antibody specifically detects ELL in Human samples. It is validated for Western Blot
Specifications
| ELL | |
| Polyclonal | |
| Western Blot | |
| C19orf17, chromosome 19 open reading frame 17, DKFZp434I1916, Eleven-nineteen lysine-rich leukemia protein, ELL gene (11-19 lysine-rich leukemia gene), ELL1, elongation factor RNA polymerase II, MEN, RNA polymerase II elongation factor ELL | |
| The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human eleven-nineteen lysine-rich leukemia using the numbering given in entry NP_006523.1 (GeneID 8178). | |
| 0.1 mL | |
| Cancer | |
| 8178 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction