missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELK4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA555994

Product Code. 15723911

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MDPMTVGRIE GDCESLNFSE VSSSSKDVEN GGKDKPPQPG AKTSSRNDYI HSGLYSSFTL NSLNSSNVKL FKLIKTENPA EKLAEKKSPQ EPTPSVIKFV TTPSKKPPVE PVAATISIGP S Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

The ELK4 gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene. The protein encoded by ELK4 is phosphorylated by the kinases, MAPK1 and MAPK8. This gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene. The protein encoded by this gene is phosphorylated by the kinases, MAPK1 and MAPK8. Several transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELK4
Polyclonal
Unconjugated
ELK4
2310011G17Rik; A130026I01Rik; BB162516; ELK4; ELK4, ETS transcription factor; ELK4, ETS-domain protein (SRF accessory protein 1); ELK4, member of ETS oncogene family; ETS domain-containing protein Elk-4; ETS domain-containing protein Elk-4-like protein; ETS transcription factor ELK4; Sap1; SAP-1; serum response factor accessory protein 1; SRF accessory protein 1
Rabbit
Antigen affinity chromatography
RUO
2005
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P28324
ELK4
Recombinant protein corresponding to Human ELK4. Recombinant protein control fragment (Product #RP-100728).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.