missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568978
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
The protein encoded by this gene is a member of the ETS-domain transcription factor family and the ternary complex factor (TCF) subfamily. Proteins in this subfamily regulate transcription when recruited by serum response factor to bind to serum response elements. This protein is activated by signal-induced phosphorylation; studies in rodents suggest that it is a transcriptional inhibitor in the absence of Ras, but activates transcription when Ras is present.
Specifications
| ELK3 | |
| Polyclonal | |
| Unconjugated | |
| ELK3 | |
| D430049E23Rik; ELK3; ELK3, ETS transcription factor; ELK3, ETS-domain protein; ELK3, ETS-domain protein (SRF accessory protein 2); ELK3, member of ETS oncogene family; Erp; Etrp; ETS domain-containing protein Elk-3; ETS transcription factor ELK3; ETS-domain protein ELK-3; ETS-related protein ERP; ETS-related protein NET; Net; SAP2; Sap-2; Serum response factor accessory protein 2; si:ch211-172b12.2; si:dkey-201e17.1; SRF accessory protein 2; zgc:198153; zgc:198183 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13713 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P41971 | |
| ELK3 | |
| synthetic peptide corresponding to a region of mouse ELK3 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction