missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1, p Ser383 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05380-20ul
This item is not returnable.
View return policy
Description
Elk-1 Polyclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western Blot
Specifications
| Elk-1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| A synthetic phosphorylated peptide around S383 of human ELK1 (NP_005220.2). TLSPI | |
| 20 μg | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific | |
| 2002 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction