missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1 Mouse anti-Human, Clone: PCRP-ELK1-1D9, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Elk-1 |
|---|---|
| Clone | PCRP-ELK1-1D9 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, Immunofluorescence, Peptide Array |
Description
Elk-1 Monoclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| Elk-1 | |
| 0.2 mg/mL | |
| Western Blot, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 2002 | |
| IgG2b | |
| Protein A or G purified |
| PCRP-ELK1-1D9 | |
| Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Recombinant full-length human Elk-1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title