missing translation for 'onlineSavingsMsg'
Learn More

Elk-1 Antibody (PCRP-ELK1-1D9), Allophycocyanin, Novus Biologicals™

Product Code. 18694017 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18694017 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18694017 Supplier Novus Biologicals Supplier No. NBP308888APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Elk-1 Monoclonal antibody specifically detects Elk-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elk-1
Applications Western Blot, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-ELK1-1D9
Conjugate APC
Formulation PBS
Gene Alias ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
Host Species Mouse
Immunogen Recombinant full-length human Elk-1 protein (Uniprot: P19419)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, MAP Kinase Signaling, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2002
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.