missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1 Antibody (3H6D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47553
This item is not returnable.
View return policy
Description
Elk-1 Monoclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Elk-1 | |
| Monoclonal | |
| Unconjugated | |
| P19419 | |
| ELK1 | |
| Purified recombinant fragment of ELK1 expressed in E. Coli. | |
| 0.1 mL | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience | |
| 2002 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3H6D12 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction