missing translation for 'onlineSavingsMsg'
Learn More

Elk-1 Antibody (3H6D12), Novus Biologicals™

Mouse Monoclonal Antibody

£187.00

Specifications

Antigen Elk-1
Clone 3H6D12
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200-1:1000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18578281
View Documents
Novus Biologicals
NBP1-47553-0.025ml
0.025 mL
£187.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Elk-1 Monoclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Elk-1
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200-1:1000
Monoclonal
Ascites
RUO
Human
ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
ELK1
IgG1
Unpurified
3H6D12
Western Blot
Unconjugated
Mouse
Autophagy, Cancer, MAP Kinase Signaling, Neuroscience
P19419
2002
Purified recombinant fragment of ELK1 expressed in E. Coli.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.