missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD ELISPOT Sets
Click to view available options
Antigen:
Granzyme B
IFN-gamma
IL-10
IL-2
IL-4
IL-5
IL-6
TNF
Cross Reactivity:
Human
Mouse
Product Type:
Human Granzyme B ELISPOT set
Human IFN-gamma ELISPOT set
Human IL-10 ELISPOT set
Human IL-2 ELISPOT set
Human IL-4 ELISPOT set
Human IL-5 ELISPOT set
Human TNF ELISPOT set
Mouse IFN-gamma ELISPOT set
Mouse IL-2 ELISPOT set
Mouse IL-4 ELISPOT set
Mouse IL-5 ELISPOT set
Mouse IL-6 ELISPOT set
Mouse TNF ELISPOT set
Unit Size:
Each
Description
Includes
Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations
- High sensitivity
- High throughput, high content analysis
- Minimal volume of biological material required
- Suitable for frozen/thawed biological samples
- Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis
Specifications
Specifications
| Antigen | IFN-gamma |
| Cross Reactivity | Human |
| Includes | Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates |
| Quantity | 10 x 96 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Cloning, proliferation assays, flow cytometry or other methods of analysis |
| Product Type | Human IFN-gamma ELISPOT set |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction