missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ELISA Systems™ Peanut (Sensitive) Residue Detection Kit for Food Allergen Testing

Product Code. 15559123
Change view
Click to view available options
Quantity:
48 Tests
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15559123 48 Tests 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15559123 Supplier Thermo Scientific™ Supplier No. ESPRDS48
  • ONLINE EXCLUSIVE PRICE
    £869.00
    £791.13 / Pack of 1
    Save £77.87
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect and quantify Ara h2 peanut allergen in food and environmental samples with this enhanced and extra sensitive assay.

Simplify allergen testing with ELISA Systems™ Peanut (Sensitive) Residue Detection Kit which helps ensure compliance with food product labeling requirements. The enzyme-linked immunosorbent assay is designed to deliver simple, sensitive, and fast detection and quantification ofpeanut protein residue in food and environmental samples. This enhanced, extra sensitive assay is developed and optimized to specifically detect Ara h2 protein.

Peanuts are a major cause of food allergies, both in children and in adults, and can induce anaphylactic shock.1 In recent years, peanut allergens have been identified and characterized. The protein Ara h2 has been one of the peanut proteins of much attention.2

Use ELISA Systems Peanut (Sensitive) Residue Detection Kit for the rapid detection and quantification of Ara h2 protein in food and environmental samples.

  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting

All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.
TRUSTED_SUSTAINABILITY

Specifications

Description ELISA Systems Peanut Residue (Sensitive) Detection Kit
Detectable Analytes Ara h 2 Peanut Alergen
Immunoassay Kit Format ELISA (Standard)
Kit Contents 20X Extraction solution concentrate, 20X Extraction Solution concentrate for samples containing polyphenols, 20X wash buffer solution concentrate, Enzyme conjugate, Negative control standard, Positive control standard, Stop solution, Substrate, Test strips
Quantity 48 Tests
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.