missing translation for 'onlineSavingsMsg'
Learn More

ELISA Systems™ Mustard Seed Protein Residue Detection kit for Food Allergen Testing

Product Code. 13235699
Change view
Click to view available options
Quantity:
48 Tests
Unit Size:
48 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13235699 48 Tests 48 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13235699 Supplier Thermo Scientific Oxoid Supplier No. ESMUS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapidly detect and quantify mustard seed protein residue in food and environmental samples.

Simplify allergen testing with ELISA Systems™ Mustard Seed Protein Residue Detection Kit which helps ensure compliance with food product labeling requirements. The enzyme-linked immunosorbent assay is designed to deliver simple, sensitive, and fast detection and quantification of mustard seed protein material in food and environmental samples. This complete, ready-to-use kit is developed and optimized for rapid and reliable test results that significantly reduce the time required to screen appropriate food products for the presence of mustard seed protein residue.

The European Directive 2003/89/EC includes mustard as a declared food allergen, and as such, it must be included, where applicable, under the appropriate labelling laws regarding food allergens. Mustard is widely used as a flavoring agent, as well as for its texture-enhancing and emulsification capacity in various food preparations, particularly with salad dressings and meats.

Use ELISA Systems Mustard Seed Protein Residue Kit for rapid detection and quantification of mustard seed protein material in food and environmental samples.

  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting


Two common and closely related species of mustard seeds were used in the development of this assay: White Mustard ( Brassica alba, sometimes known as Sinapis alba or Yellow Mustard) and Black Mustard ( Brassica nigra). The assay has been shown to detect the seed proteins from these species, as well as seed protein from another mustard seed: Brassica juncea (Brown Mustard).

All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.
TRUSTED_SUSTAINABILITY

Specifications

Description ELISA Systems Mustard Seed Protein Residue Detection Kit
Detectable Analytes Mustard Seed Protein Residue
Immunoassay Kit Format ELISA (Standard)
Product Type Mustard Seed Protein Residue Detection Kit
Kit Contents 20X wash buffer solution concentrate, Enzyme conjugate, Negative Standard, Positive Standards, Stop solution, Substrate, Test strips
Quantity 48 Tests
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.