missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ELISA Systems™ Lupin Residue Detection Kit for Food Allergen Testing

Product Code. 13225699
Change view
Click to view available options
Quantity:
48 Tests
Unit Size:
48 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13225699 48 Tests 48 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13225699 Supplier Thermo Scientific™ Supplier No. ESLFP48

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rapidly detect and quantify lupin residues in food and environmental samples.

Simplify allergen testing with ELISA Systems™ Lupin Residue Detection Kit which helps ensure compliance with food product labeling requirements. The enzyme-linked immunosorbent assay is designed to deliver simple, sensitive, and fast detection and quantification of lupin residue in food and environmental samples. This complete, ready-to-use kit is developed and optimized for rapid and reliable test results that significantly reduce the time required to screen appropriate food products for the presence of lupin residues.

Lupin refers to a range of plants in the genus Lupinus which are members of the legume (Fabaceae) family. Lupins have been commonly used as food throughout Europe since the Roman Age. In recent years, lupin flour has been produced from lupin seeds and is becoming increasingly popular as an ingredient in a wide variety of foods as an alternative to wheat flour.The most commonly cultivated lupins for food use are the white lupin (L. albus), the blue lupin (L. Angustifolius), and the yellow lupin (L. Luteus).

Use ELISA Systems Lupin Residue Kit for rapid detection and quantification of lupin material in food and environmental samples.

  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting


The incidence of lupin allergy is significantly higher in individuals with peanut allergy1 but may also be present in individuals with no sensitivity to peanut2, 3. Allergic reaction to lupin ranges from oral allergy syndrome to severe anaphylactic reactions.

Lupin and derived products are declarable allergens in the EU according to directive 2006/142/EC.

This kit detects the three most common species used in food production: L. albus, L. angustifolius, and L. luteus.

All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.
TRUSTED_SUSTAINABILITY

Specifications

Description ELISA Systems Lupin Residue Detection Kit
Detectable Analytes Lupin residue
Immunoassay Kit Format ELISA (Standard)
Product Type Lupin Residue Detection Kit
Kit Contents 20X Extraction solution concentrate, 20X wash buffer solution concentrate, Enzyme conjugate, Negative Standard, Positive Standards, Stop solution, Substrate, Test strips
Quantity 48 Tests
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.