missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ELISA Systems™ Food Allergen Assay Accessories
Click to view available options
Product Type:
Extraction Solution (Polyphenol)
Unit Size:
25mL
Description
Support the functionality of ELISA Systems™ Food Allergen Test Kits with ELISA Systems Food Allergen Assay Accessories, including extraction and wash solutions. ELISA Systems Food Allergen Test Kits are a comprehensive range of enzyme-linked immunosorbent assays (ELISA), designed to detect and quantify the presence of specific proteins or residues in food products and environmental samples, to help you ensure compliance with food product labeling requirements.
ELISA Systems Food Allergen Test Kits come complete and ready-to-use. Solutions can be ordered separately, where required:
- ELISA Systems Extraction Solution: Compatible with most of the ELISA Systems Food Allergen Test Kits, thereby reducing sample preparation time
- ELISA Systems Extraction Solution Polyphenol: A special extraction solution for samples containing polyphenols, including dark chocolate, wine, fruit juices, herbs, and tannins. Already included with nut test kits
- ELISA Systems Wash Solution
- Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
- Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
- Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
- Flexible: Use for qualitative or quantitative testing—a single kit for more applications
- Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
- Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting
Specifications
Specifications
| Concentration | 20X |
| Description | ELISA Systems Extraction Solution Polyphenol (20X concentrate) |
| Immunoassay Kit Format | ELISA (Standard) |
| Product Type | Extraction Solution (Polyphenol) |
| Quantity | 25 mL Bottle |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur