missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ELISA/ELISPOT Diluent (5X)

Product Code. 12760000
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12760000 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12760000 Supplier Invitrogen™ Supplier No. 00420256
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Supplemented with fetal bovine serum as source of animal serum proteins. Used for blocking non-specific binding of ELISA plates, diluting recombinant standards and test samples and making the working dilutions of secondary detection reagent. Also contains 1% ProClin 150 as a preservative

The 5X ELISA/ELISPOT Diluent is a Phosphate Buffered Solution supplemented with fetal bovine serum as a source of animal serum proteins. This buffer is used for blocking non-specific binding of ELISA plates, diluting recombinant standards and test samples, and making the working dilutions of the secondary detection reagent (i.e., detection antibody-biotin and AV-HRP Cat. No. 18-4100). This diluent solution also contains a preservative.

Reported Application
ELISA

TRUSTED_SUSTAINABILITY

Specifications

Product Type Diluent
Quantity 100 mL
Form Liquid
For Use With (Application) ELISA
Product Line eBioscience

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.