missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA/ELISPOT Coating Buffer Powder

Product Code. 15556266
Click to view available options
Quantity:
1 each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15556266

Brand: Invitrogen™ 00004459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed and tested for use as the working solution for coating the ELISA or ELISPOT capture antibody to the ELISA or ELISPOT plate microwells. This has been tested with all cytokine ELISA and ELISPOT capture antibodies, according to the procedures described in the corresponding protocols.

  • Dried powder formulation of phosphate buffered saline
  • Each packet is reconstituted in 1 Liter of distilled, deionized water, then filtered using a 0.22μm filter
  • Resulting 1x PBS (pH 7.4) is suitable for use as the ELISA/ELISPOT coating buffer
TRUSTED_SUSTAINABILITY

Specifications

Product Type Coating Buffer
Quantity 1 each
Form Powder
For Use With (Application) ELISA, ELISPOT

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.