missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems ELISA Buffer Set-96T

Product Code. 17196271
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests

Product Code. 17196271

Brand: ACROBiosystems EBS00196tests

Please to purchase this item. Need a web account? Register with us today!

The ELISA Buffer Set-96T can be quickly applied to the ELISA experiment and save your time to prepare buffers

Specifications

Product Type buffer kits
Kit Contents This ELISA Buffer Set-96 T contains a High-bind 96 well plate, coating buffer, washing buffer, blocking buffer, substrate solution and stop solution. The ELISA Buffer Set-96 T can be quickly applied to the ELISA experiment and save your time to prepare buffe
Quantity 96 Tests
Shipping Condition RT
Storage Requirements 2°C to 8°C, aviod light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.