missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems ELISA Assay Kit for Anti-PD-1 h-mAb in Mouse Serum

Product Code. 15806677
Click to view available options
Quantity:
96 Tests
Unit Size:
1 set

Product Code. 15806677

Brand: ACROBiosystems EPMV196TESTS

Please to purchase this item. Need a web account? Register with us today!

For quantification of monoclonal anti-PD-1 antibodies in mouse serum.

How a therapeutic antibody is metabolized in the body is pertinently relevant to its efficacy. Therefore, pharmacokinetics study is an important part of the drug development. PD-1 is probably the most targeted molecule in today’s pharmaceutical industry, thanking to the clinical success of Keytruda® and Opdivo®. Many investigational anti-PD-1 mAbs are being developed. There is a growing need for a standard assay that can be used to facilitate the study of their pharmacokinetics.

Specifications

Product Type Pk. Assay Kit
Kit Contents The enclosed ELISA assay kit for anti-PD-1 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant PD-1 protein (ECD) and a biotinylated anti-PD-1 monoclonal antibody. The method employs the principle of competitive ELISA, allow
Quantity 96 Tests
Shipping Condition RT
Storage Requirements -20°C, aviod light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.