missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems ELISA Assay Kit for Anti-PD-1 h-mAb in Monkey Serum
Click to view available options
Quantity:
96 Tests
Unit Size:
1 set
Description
How a therapeutic antibody is metabolized in the body is pertinently relevant to its efficacy. Therefore, pharmacokinetics study is an important part of the drug development. PD-1 is probably the most targeted molecule in today’s pharmaceutical industry, thanking to the clinical success of Keytruda® and Opdivo®. Many investigational anti-PD-1 mAbs are being developed. There is a growing need for a standard assay that can be used to facilitate the study of their pharmacokinetics.
Specifications
Specifications
| Product Type | Pk. Assay Kit |
| Kit Contents | The enclosed ELISA assay kit for anti-PD-1 h-mAb in monkey serum is based on an enzyme immunoassay (ELISA) between recombinant PD-1 protein (ECD) and a biotinylated anti-PD-1 monoclonal antibody. The method employs the principle of competitive ELISA, allo |
| Quantity | 96 Tests |
| Shipping Condition | RT |
| Storage Requirements | -20°C, aviod light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction