missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems ELISA Assay Kit for Anti-HER-2 h-mAb in Monkey Serum

Product Code. 15846637
Click to view available options
Quantity:
480 Tests
Unit Size:
1 set

Product Code. 15846637

Brand: ACROBiosystems EHCV1480TESTS

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

For quantification of monoclonal anti-HER-2 antibodies in monkey serum.

How a therapeutic antibody is metabolized in the body is pertinently relevant to its efficacy. Therefore, pharmacokinetics study is an important part of the drug development. HER-2 is probably the most targeted molecule in today’s pharmaceutical industry, thanking to the clinical success of Herceptin®. Many investigational anti-HER-2 mAbs are being developed. There is a growing need for a standard assay that can be used to facilitate the study of their pharmacokinetics.

Specifications

Product Type Pk. Assay Kit
Kit Contents The enclosed ELISA assay kit for anti-HER-2 h-mAb in monkey serum is based on an enzyme immunoassay (ELISA) between recombinant HER-2 protein (ECD) and a biotinylated anti-HER-2 monoclonal antibody. The method employs the principle of competitive ELISA, a
Quantity 480 Tests
Shipping Condition RT
Storage Requirements -20°C, aviod light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.