missing translation for 'onlineSavingsMsg'
Learn More

Elimusertib, MedChemExpress

Product Code. 16463418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463418 10 mg 10mg
16442766 25 mg 25mg
16434646 100 mg 100mg
16426147 5 mg 5mg
16419946 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463418 Supplier Medchem Express Supplier No. HY101566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elimusertib (BAY-1895344) is a potent, orally active and selective ATR inhibitor with an IC50 of 7 nM. Elimusertib has anti-tumor activity. Elimusertib can be used for the research of solid tumors and lymphomas.

Elimusertib (BAY-1895344) is a potent, orally active and selective ATR inhibitor with an IC50 of 7 nM. Elimusertib has anti-tumor activity. Elimusertib can be used for the research of solid tumors and lymphomas.

Specifications

Chemical Name or Material Elimusertib
CAS 1876467-74-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H21N7O
Quantity 10 mg
Synonym BAY 1895344
Solubility Information DMSO : 5.4 mg/mL (14.38 mM; Need ultrasonic)
SMILES CN1N=CC=C1C2=C(C=CN=C3C4=CC=NN4)C3=NC(N5[C@H](C)COCC5)=C2
Molecular Weight (g/mol) 375.43
Formula Weight 375.43
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.