missing translation for 'onlineSavingsMsg'
Learn More

Elimusertib hydrochloride, MedChemExpress

Product Code. 16473418
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473418

Brand: Medchem Express HY101566A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elimusertib (BAY 1895344) hydrochloride is a potent, orally active and selective ATR inhibitor with an IC50 of 7 nM. Elimusertib hydrochloride has anti-tumor activity. Elimusertib hydrochloride can be used for the research of solid tumors and lymphomas.

Elimusertib (BAY 1895344) hydrochloride is a potent, orally active and selective ATR inhibitor with an IC50 of 7 nM. Elimusertib hydrochloride has anti-tumor activity. Elimusertib hydrochloride can be used for the research of solid tumors and lymphomas.

Specifications

Chemical Name or Material Elimusertib hydrochloride
Color Orange
Purity Grade Notes Research
Molecular Formula C20H22ClN7O
Quantity 10 mg
Synonym BAY 1895344 hydrochloride
Solubility Information DMSO : 50 mg/mL (121.39 mM; Need ultrasonic) ∣H2O : 50 mg/mL (121.39 mM; Need ultrasonic)
SMILES CN1N=CC=C1C2=C(C=CN=C3C4=CC=NN4)C3=NC(N5[C@H](C)COCC5)=C2.[H]Cl
Molecular Weight (g/mol) 411.89
Formula Weight 411.89
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.