missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELFN1 Monoclonal Antibody (965705)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560901
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Postsynaptic protein that regulates circuit dynamics in the central nervous system by modulating the temporal dynamics of interneuron recruitment. Specifically present in excitatory synapses onto oriens-lacunosum molecular (OLM) interneurons and acts as a regulator of presynaptic release probability to direct the formation of highly facilitating pyramidal-OLM synapses. Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
Specifications
| ELFN1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P0C7U0 | |
| ELFN1 | |
| Chinese Hamster Ovary cell line, CHO-derived human ELFN1 protein, Asp28-Tyr418. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 965705 | |
| Unconjugated | |
| ELFN1 | |
| ELFN1; extracellular leucine rich repeat and fibronectin type III domain containing 1; extracellular leucine-rich repeat and fibronectin type III containing 1; extracellular leucine-rich repeat and fibronectin type III domain containing 1; extracellular leucine-rich repeat and fibronectin type-III domain-containing protein 1; PPP1R28; Protein ELFN1; protein phosphatase 1 regulatory subunit 28; protein phosphatase 1, regulatory subunit 28 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 392617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction