missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568805
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 85%; Horse: 92%; Human: 92%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%.
ELF5 is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus.
Specifications
| ELF5 | |
| Polyclonal | |
| Unconjugated | |
| ELF5 | |
| E74 like ETS transcription factor 5; E74-like factor 5; E74-like factor 5 (ets domain transcription factor); E74-like factor 5 ESE-2a; Elf5; elf-5; Epithelium specific Ets-2; epithelium-restricted ESE-1-related Ets factor; Epithelium-specific Ets transcription factor 2; ESE2; ESE-2; ESE-5; ESE-5.; ets transcription factor; ETS-related transcription factor Elf-5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13711 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8VDK3 | |
| ELF5 | |
| synthetic peptide directed towards the middle region of mouse ELF5. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction