missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568837
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%; Rat: 85%.
ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5'-WGGA-3'. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
Specifications
| ELF4 | |
| Polyclonal | |
| Unconjugated | |
| Elf4 | |
| AV314029; BC042423; E74 like ETS transcription factor 4; E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ELFR; ENSMUSG00000053512; ETS-related transcription factor Elf-4; Gm9907; MEF; myeloid elf-1 like factor; myeloid Elf-1-like factor; RGD1560743 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56501 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Z2U4 | |
| Elf4 | |
| synthetic peptide corresponding to a region of mouse ELF4 (aa 141-190). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction